
Iron(III) oxide ≥95%, powder, Reagent Grade
Supplier: Spectrum Chemical
New Product
Certificates
About this item
CAS Number:1309-37-1
:
Formula:Fe₂O₃
MDL Number:MFCD00011008
:159.69 g/mol
Merck Index:13,04058
Melting Pt:1538 °C
Density:5.24 g/cm³ (20 °C)
EINECS:215-168-2
:GHS07
Storage Temperature:Ambient