Order Entry
Sweden
ContactUsLinkComponent
Hydrochloric acid solution 103 g/L
Hydrochloric acid solution 103 g/L
Catalog # REGEHSP1031
Supplier:  REAGECON
New Product
CAS Number:  
Hydrochloric acid solution 103 g/L
Catalog # REGEHSP1031
Supplier:  REAGECON
New Product
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle GL45
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
ADR:8,II
 :36.46 g/mol
UN:1789
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324