Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Isoflucypram
Isoflucypram
Isoflucypram
Catalog # EHERC14424700
Supplier:
EHRENSTORFER
New Product
CAS Number:
Expand image
Isoflucypram
Catalog # EHERC14424700
Supplier:
EHRENSTORFER
New Product
CAS Number:
Specifications
Pk:
50 mg
CAS No.:
1255734-28-1
Description:
Isoflucypram
Formula:
C₁₉H₂₁ClF₃N₃O
Storage temperature:
20 °C
Molecular weight (Mw):
399.84
Specifications
About this item
Product Details
Product Details