Specifications
- Pk:100 mg
- CAS No.:67564-91-4
- Description:Fenpropimorph
- Storage conditions:–20 °C
- Hazard class description:H302 - H315 - H361d - H411
- Shipping conditions:WICE
- MDL No.:MFCD00144306
- M-Clarity quality level:MQ300
- Molecular weight:303,48
- Flash point:>100 °C - closed cup
Specifications
About this item
Fenpropimorph is a systematic fungicide that belongs to the morpholine group of sterol biosynthesis inhibitors. It is widely applied in cereals to control pathogens such as Erysiphe graminis (powdery mildew) and Puccinia recondita (cereal rust). Fenpropimorph inhibits fungal; 8 to 7 isomerase and 14 reductase enzymes involved in the ergosterol biosynthesis. Fenpropimorph is not approved for use in European Union (EU) as per Regulation (EC) No 1107/2009. The maximum residue level (MRLs) for fenpropimorph (sum of isomers) following Reg. (EU) 2019/552 has been set from 0,01 to 3 mg/kg for products of plant and animal origin. Fenpropimorph has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP) run within the EU and EFTA in/on products of plant origin.