Order Entry
Sweden
ContactUsLinkComponent
Supelco® Organic Reference Standard, F.A.M.E. Mix C14 - C22
Supelco® Organic Reference Standard, F.A.M.E. Mix C14 - C22
  CRM18917
 :  Merck
 :  
Supelco® Organic Reference Standard, F.A.M.E. Mix C14 - C22
  CRM18917
 :  Merck
 :  

 

  • Concentration:
    Wt.% (varied conc.)
  • Pk:
    100 mg
  • Standards:
    Certified reference material
  • Description:
    F.A.M.E. mix C14 - C22
  • Storage conditions:
    –10...–25 °C
  • Hazard class description:
    H413
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Pk Info:
    Ampule of 100 mg

 

 

This certified reference material is useful for determining relative retention times and approximating response factors.

 : P273 - P501