Supelco® TraceCERT® Organic Reference Standard, Isoxaben
Isoxaben, Certified reference material, TraceCERT®, 50 mg
Catalog # 75772-50MG
Supplier: Merck
Supelco® TraceCERT® Organic Reference Standard, Isoxaben
Catalog # 75772-50MG
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Isoxaben CRM may also find its use as described below:
- To measure the effect of isoxaben on the incorporation of radiolabeled glucose leucine uracil and acetate into acid-insoluble cell wall material protein nucleic acids and fatty acids respectively in Arabidopsis thaliana
- To determine the relative toxicities of isoxaben and two of its primary metabolites in soybean and wheat cell cultures
- To investigate the effect of trifluralin pronamide haloxyfop-p methyl propaquizafop and isoxaben on weed control and oilseed rape yield
- To study the degradation of isoxaben in soil and an aqueous system followed by isolation and identification of degradation products formed
- To examine the effect of four different organic wastes composted sheep manure spent coffee grounds composted pine bark and coir on potential groundwater pollution of propanil isoxaben cadusafos and pencycuron under laboratory conditions
Caution: P273 - P501
Specifications
Specifications
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- Description:Isoxaben
- Cat. No.:75772-50MG
- M-Clarity quality level:MQ300
- Storage conditions:2…8 °C
- Hazard class description:H413
- MDL No.:MFCD00078687
- Molecular weight:332,39
- CAS No.:82558-50-7
- Shipping conditions:STD
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



