Skip to Header
Skip to Main Content
Skip to Footer
Avancerad
Avancerad
SDS
Certifikat
Tillverkardata
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Digoxigenin (DIG) alkyne
Digoxigenin (DIG) alkyne, FastClick™
Digoxigenin (DIG) alkyne, FastClick™
Digoxigenin (DIG) alkyne, FastClick™
Catalog # AATB72900
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
Digoxigenin (DIG) alkyne, FastClick™
Catalog # AATB72900
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Produktinformation
:
:
870.15 g/mol
Produktinformation