Anti-NT5C1B Rabbit Polyclonal Antibody
ANTIA100477-100
New Product
- Antibody type:Primary
- Antigen name:5' nucleotidase cytosolic IB
- Antigen symbol:NT5C1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q96P26
- Antigen synonyms:cN1B|cN-IB|CN IB|Cytosolic 5' nucleotidase 1B|5NT1B_HUMAN|Autoimmune infertility-related protein|AIRP|5' nucleotidase cytosolic IB isoform CRA_b|CNIB
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:68 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human NT5C1B (amino acids 171 - 220)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to NT5C1B for IF and ELISA with samples derived from human.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: NT5C1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human