Order Entry
Sweden
ContactUsLinkComponent
Anti-D6 Rabbit Polyclonal Antibody
Anti-D6 Rabbit Polyclonal Antibody
  ANTIA101186-100
New Product
undefined
Anti-D6 Rabbit Polyclonal Antibody
  ANTIA101186-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ACKR2
  • Antigen symbol:
    D6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O00590
  • Antigen synonyms:
    CCBP2|CCR10|Chemokine receptor CCR 10|Chemokine binding protein 2|Atypical chemokine receptor 2|CMKBR9|CCR9|Chemokine receptor CCR 9|C C chemokine receptor D6
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    43 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human CCBP2 (amino acids 334-383)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to CCBP2 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: D6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human