Order Entry
Sweden
ContactUsLinkComponent
Anti-GPCR MCH2 Rabbit Polyclonal Antibody
Anti-GPCR MCH2 Rabbit Polyclonal Antibody
  ANTIA100988-100
New Product
undefined
Anti-GPCR MCH2 Rabbit Polyclonal Antibody
  ANTIA100988-100
New Product

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    G protein coupled receptor 145
  • Antigensymbol:
    GPCR MCH2
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gen-ID:
    UniprotID# Q969V1
  • Antigen synonymer:
    G-protein coupled receptor 145|MCH2R|GPR145|MCH-R2|MCH-2R|G protein coupled receptor slt|GPCR MCH2|MCH receptor 2|GPRv17
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molekylvikt:
    38 kDa
  • Förvaringstemperatur:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human MCHR2 (amino acids 291-340)
  • Rening:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Förpackningstyp:
    Plastic vial
  • Förp.:
    100 µG

 

 

Rabbit polyclonal antibody to MCHR2 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000

Type: Primary
Antigen: GPCR MCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human