Anti-Rad51D Rabbit Polyclonal Antibody
Catalog # ANTIA100119-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:BROVCA4
- Antigen symbol:Rad51D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O75771
- Antigen synonyms:HsTRAD|OTTHUMP00000163853|DNA repair protein RAD51 homolog 4|RA51D_HUMAN|OTTHUMP00000163851|RAD51 homolog D (S. cerevisiae)|R51H3|RAD51 homolog D|OTTHUMP00000163852
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:35 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human RAD51L3 (amino acids 131-180)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to RAD51L3 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: Rad51D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human