Order Entry
Sweden
ContactUsLinkComponent
Anti-Factor B Rabbit Polyclonal Antibody
Anti-Factor B Rabbit Polyclonal Antibody
Catalog # ANTIA94484-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Factor B Rabbit Polyclonal Antibody
Catalog # ANTIA94484-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    AHUS4
  • Antigensymbol:
    Factor B
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gen-ID:
    UniprotID# P00751
  • Antigen synonymer:
    BFD|CFAB|B factor properdin|CFAB_HUMAN|C3 proactivator|BF|C3/C5 convertase|CFB|C3 proaccelerator
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molekylvikt:
    57 and 85 kDa
  • Förvaringstemperatur:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human CFAB Bb (amino acids 241-290)
  • Rening:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
  • Förpackningstyp:
    Plastic vial
  • Förp.:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to CFAB Bb (cleaved Lys260) for WB and ELISA with samples derived from Human and Mouse.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:10000

Type: Primary
Antigen: Factor B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse