Order Entry
Sweden
ContactUsLinkComponent
 
Anti-Cytochrome P450 2E1 Rabbit Polyclonal Antibody
  ANTIA93565-100
New Product
undefined
Anti-Cytochrome P450 2E1 Rabbit Polyclonal Antibody
  ANTIA93565-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    4-nitrophenol 2-hydroxylase
  • Antigen symbol:
    Cytochrome P450 2E1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05181
  • Antigen synonyms:
    Cytochrome P450 isozyme 3A|CYPIIE1|Cytochrome P450 2E1|CYP2E|CPE1|CP2E1_HUMAN|Cytochrome P450, family 2, subfamily e, polypeptide 1|Cyp2e1|Cytochrome P450 LM3A
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    56 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human Cytochrome P450 2E1 (amino acids 371-420)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to Cytochrome P450 2E1 for IHC, IF and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: IHC, IF, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:1000

Type: Primary
Antigen: Cytochrome P450 2E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat