Anti-pro Caspase 3 Rabbit Polyclonal Antibody
ANTIA93475-100
New Product
- Antibody type:Primary
- Antigen name:CASP3
- Antigen symbol:pro Caspase 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P42574
- Antigen synonyms:Yama|pro Caspase 3|Caspase 3 apoptosis related cysteine peptidase|CPP32|SCA1|CPP32B|Procaspase 3|SCA 1|Caspase 3
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:31 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Caspase 3 around the phosphorylation site of Ser150 (amino acids 116-165)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to Caspase 3 (phospho Ser150) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: pro Caspase 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat