Anti-Blood Group A1B Antigen Mouse Monoclonal Antibody [Clone: HE-24]
Catalog # ANTIA86814-1
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Blood Group A1B Antigen
- Antigen symbol:Blood Group A1B Antigen
- Clonality:Monoclonal
- Clone:HE-24
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Form:Liquid
- Storage buffer:Supplied as an aliquot of concentrated tissue culture supernatant
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Glycoprotein fraction isolated from saliva of secretors with blood group A
- Tested applications:Agglutination
- Purification:Hybridoma culture supernatant is 4X concentrated by ultrafiltration using 100 kDa cut off membrane
- Pack type:Plastic vial
- Pk:1 mL
Specifications
About this item
Mouse monoclonal [HE-24] antibody to Blood Group A1B Antigen for AGG with samples derived from Human.
- Validated applications: AGG
Type: Primary
Antigen: Blood Group A1B Antigen
Clonality: Monoclonal
Clone: HE-24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human