Anti-CD88 Mouse Monoclonal Antibody [Clone: S5/1]
ANTIA86678-100
New Product
- Antibody type:Primary
- Antigen name:ANAPHYLATOXIN RECEPTOR
- Antigen symbol:CD88
- Clonality:Monoclonal
- Clone:S5/1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Rabbit,Ferret,Human,Bovine
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P21730
- Antigen synonyms:C5a anaphylatoxin chemotactic receptor|C5a-R|c5ar1|C5aR|C5R1|C5a anaphylatoxin receptor|C5A|C5a ligand|C5AR_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant N-terminal peptide (Asp15-Asp27) of human C5aR
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [S5/1] antibody to CD88 for Flow Cytometry, IP, WB and IHC-P with samples derived from Human, Bovine, Ferret and Rabbit.
- Validated applications: Flow cytometry, IP, WB, IHC-P
- Recommended dilutions: Flow cytometry: 3 to 12 µg/ml
Type: Primary
Antigen: CD88
Clonality: Monoclonal
Clone: S5/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Bovine, Ferret, Rabbit