Order Entry
Sweden
ContactUsLinkComponent
Anti-NKG2D Mouse Monoclonal Antibody [Clone: 1D11]
Anti-NKG2D Mouse Monoclonal Antibody [Clone: 1D11]
  ANTIA86459-100
New Product
Anti-NKG2D Mouse Monoclonal Antibody [Clone: 1D11]
  ANTIA86459-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CD314
  • Antigen symbol:
    NKG2D
  • Clonality:
    Monoclonal
  • Clone:
    1D11
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P26718
  • Antigen synonyms:
    KLRK1|NK cell receptor D|Killer cell lectin like receptor subfamily K member 1|KLR|NK lectin-like receptor|Killer cell lectin-like receptor subfamily K member 1|KLRC4 KLRK1 readthrough|CD314 antigen|D12S2489E
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    NKL cell line
  • Tested applications:
    IHC-Fr
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [1D11] antibody to NKG2D for Flow Cytometry, IP and IHC-Fr with samples derived from Human.

  • Validated applications: Flow cytometry, IP, IHC-Fr
  • Recommended dilutions: Flow cytometry: 1 to 4 µg/ml

Type: Primary
Antigen: NKG2D
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human