Anti-CD103 Mouse Monoclonal Antibody [Clone: Ber-ACT8] (APC (Allophycocyanin))
Catalog # ANTIA86442-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:A530055J10
- Antigen symbol:CD103
- Clonality:Monoclonal
- Clone:Ber-ACT8
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P38570
- Antigen synonyms:alpha-M290|aM290|alpha-E1|HML 1 antigen|CD 103|Antigen CD103|HML-1 antigen|CD103 antigen|HML1 antigen
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:HTLV-1 induced human T cell line MAPS16
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [Ber-ACT8] antibody to CD103 (APC) for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: Ber-ACT8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates