Anti-PAG1 Mouse Monoclonal Antibody [Clone: MEM-255]
Katalog # ANTIA86214-100
Leverantör: ANTIBODIES.COM
New Product
Specifikationer
- Antibody type:Primary
- Antigen name:20S PROTEASOME ALPHA SUBUNIT G1
- Antigen symbol:PAG1
- Clonality:Monoclonal
- Clone:MEM-255
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q3U1F9
- Antigen synonyms:20S PROTEASOME ALPHA SUBUNIT G1, 20S|T20P8.7|20S PROTEASOME SUBUNIT PAG1|T20P8_7|AT2G27020
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant intracellular fragment (aa 97-432) of human Cbp (PAG)
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifikationer
Om denna produkt
Mouse monoclonal [MEM-255] antibody to PAG1 for Flow Cytometry, WB and IHC-P with samples derived from Human.
- Validated applications: Flow cytometry, WB, IHC-P
- Recommended dilutions: Flow cytometry: 2 µg/ml
Type: Primary
Antigen: PAG1
Clonality: Monoclonal
Clone: MEM-255
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human