Order Entry
Sweden
ContactUsLinkComponent
 
Anti-Pit1 Rabbit Polyclonal Antibody
  ANTIA93298-100
New Product
undefined
Anti-Pit1 Rabbit Polyclonal Antibody
  ANTIA93298-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    GHF 1A
  • Antigen symbol:
    Pit1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28069
  • Antigen synonyms:
    GHF-1|Pit 1|Hmp 1|GHF1A|Hmp1|Dwarf|GHF1|Growth hormone factor 1
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    35 kDa
  • Sequence:
    LTPCLYKFPDHTLSHGFPPIHQPLLAEDPTAADFKQELRRKSKLVEEPIDMDSPEIRELEKFANEFKVRRIKLGYTQTNVGEALAAVHGSEFSQTTICRFENLQLSFKNACKLKAILSKWLEEAEQVGALYNEKVGANERKRKRRTTISIAAKDALERHFGEQNKPSSQEI
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 100-270 of human POU1F1 (NP_001116229)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to Pit1 for WB with samples derived from Human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:100 to 1:500

Type: Primary
Antigen: Pit1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human