Anti-NXN/NRX Rabbit Polyclonal Antibody
Catalog # ANTIA93282-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:NXN/NRX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q6DKJ4
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
- Molecular weight:48 kDa
- Sequence:CRSLTRVLVESYRKIKEAGQNFEIIFVSADRSEESFKQYFSEMPWLAVPYTDEARRSRLNRLYGIQGIPTLIMLDPQGEVITRQGRVEVLNDEDCREFPWHPKPVLELSDSNAAQLNEGPCLVLFVDSEDDGESEAAKQLIQPIAEKIIAKYKAKEEEAPLLFFVAGEDDMTDSLRDYTNL
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 100-280 of human NXN (NP_001192248.1)
- Tested applications:ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to NXN/NRX for WB and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated applications: WB, ICC/IF
- Recommended dilutions: WB: 1:500 to 1:2,000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: NXN/NRX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat