Anti-p57 Kip2 Rabbit Recombinant Antibody [clone: KIP2/7083R]
Catalog # ANTIA278541-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Beckwith Wiedemann syndrome
- Antigen symbol:p57 Kip2
- Clonality:Recombinant
- Clone:KIP2/7083R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:UniprotID# P49918
- Antigen synonyms:CDKI|CDKN1C|BWCR|CDN1C_HUMAN|Cyclin-dependent kinase inhibitor 1C|CDKN 1C|BWS|Cyclin dependent kinase inhibitor 1C|Cyclin dependent kinase inhibitor p57
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:57 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human p57 Kip2 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [KIP2/7083R] antibody to p57 Kip2 for IHC-P with samples derived from human and mouse.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 0,25 to 0,5 µg/ml
Type: Primary
Antigen: p57 Kip2
Clonality: Recombinant
Clone: KIP2/7083R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse