Anti-Apolipoprotein B Mouse Monoclonal Antibody [clone: APOB/4335]
ANTIA278238-100
New Product
- Antibody type:Primary
- Antigen name:Apo B-100
- Antigen symbol:Apolipoprotein B
- Clonality:Monoclonal
- Clone:APOB/4335
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:Multiple
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P04114
- Antigen synonyms:Apolipoprotein B-48|Apolipoprotein B (including Ag(x) antigen)|Apo B-48|ApoB 48|Apolipoprotein B|APOB|ApoB 100 |Apolipoprotein B 100|APOB_HUMAN
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:512 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 592-689, of human Apolipoprotein B. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [APOB/4335] antibody to Apolipoprotein B for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Apolipoprotein B
Clonality: Monoclonal
Clone: APOB/4335
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human