Anti-Blood Group Antigen B Mouse Monoclonal Antibody [clone: HEB-29]
Catalog # ANTIA254322-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Blood Group Antigen B
- Antigen symbol:Blood Group Antigen B
- Clonality:Monoclonal
- Clone:HEB-29
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P16442
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Mixture of erythrocytes of group B and glyco protein fraction isolated from saliva of secretors with blood group B.
- Tested applications:Agglutination
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [HEB-29] antibody to Blood Group Antigen B for ELISA, Agglutination and IF with samples derived from human.
- Validated Applications: ELISA, Agglutination, IF
- Recommended Dilutions: IF: 1 to 2 µg/ml
Type: Primary
Antigen: Blood Group Antigen B
Clonality: Monoclonal
Clone: HEB-29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human