Anti-CD1c Mouse Monoclonal Antibody [clone: CD1C/1603]
Katalog # ANTIA253659-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:BDCA1
- Antigen symbol:CD1c
- Clonality:Monoclonal
- Clone:CD1C/1603
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P29017
- Antigen synonyms:CD1C antigen c polypeptide|CD1c antigen|CD1c molecule|CD1A|Cortical thymocyte antigen CD1C|Differentiation antigen CD1 alpha 3|CD1|CD1c|CD1C_HUMAN
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:43 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CD1c protein.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [CD1C/1603] antibody to CD1c for ELISA with samples derived from human.
- Validated Applications: ELISA
Type: Primary
Antigen: CD1c
Clonality: Monoclonal
Clone: CD1C/1603
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human