Order Entry
Sweden
ContactUsLinkComponent
Anti-CD1a Mouse Monoclonal Antibody [clone: 66IIC7]
Anti-CD1a Mouse Monoclonal Antibody [clone: 66IIC7]
Catalog # ANTIA253651-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CD1a Mouse Monoclonal Antibody [clone: 66IIC7]
Catalog # ANTIA253651-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD 1a
  • Antigen symbol:
    CD1a
  • Clonality:
    Monoclonal
  • Clone:
    66IIC7
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P06126
  • Antigen synonyms:
    cortical thymocyte antigen CD1A|cluster of differentiation 1 A|CD1a|CD1|CD1A antigen, a polypeptide|CD1a molecule|differentiation antigen CD1 alpha 3|CD1A_HUMAN|CD1A Antigen
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    49 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human thymocytes.
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [66IIC7] antibody to CD1a for Flow Cytometry and IF with samples derived from human.

  • Validated Applications: Flow Cytometry, IF
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: CD1a
Clonality: Monoclonal
Clone: 66IIC7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human