Order Entry
Sweden
ContactUsLinkComponent
Anti-CD48 Rabbit Recombinant Antibody [clone: DM43] (Biotin)
Anti-CD48 Rabbit Recombinant Antibody [clone: DM43] (Biotin)
Supplier:  ANTIBODIES.COM
New Product
Certificates
Anti-CD48 Rabbit Recombinant Antibody [clone: DM43] (Biotin)
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Catalog No:
  • ANTIA317421-500
  • ANTIA317421-100
  • Antigen symbol:
  • CD48
  • CD48
  • Antigen name:
  • Antigen CD48
  • Antigen CD48
  • Antigen synonyms:
  • BCM1 surface antigen|B cell membrane protein|BLAST|BLAST1|BCM 1 surface antigen|BCM1|B lymphocyte activation marker BLAST 1|B-lymphocyte activation marker BLAST-1|B-cell activation marker
  • BCM1 surface antigen|B cell membrane protein|BLAST|BLAST1|BCM 1 surface antigen|BCM1|B lymphocyte activation marker BLAST 1|B-lymphocyte activation marker BLAST-1|B-cell activation marker
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Recombinant
  • Recombinant
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • DM43
  • DM43
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • UniprotID# P09326
  • UniprotID# P09326
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Form:
  • Lyophilised
  • Lyophilised
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Concentration:
  • Reconstitution dependent
  • Reconstitution dependent
  • Storage buffer:
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Storage temperature:
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C

Specifications