Order Entry
Sweden
ContactUsLinkComponent
Anti-KLF12 Mouse Monoclonal Antibody [Clone: PCRP-KLF12-1E3]
Anti-KLF12 Mouse Monoclonal Antibody [Clone: PCRP-KLF12-1E3]
  ANTIA277541-100
New Product
undefined
Anti-KLF12 Mouse Monoclonal Antibody [Clone: PCRP-KLF12-1E3]
  ANTIA277541-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    2700063E05Rik
  • Antigen symbol:
    KLF12
  • Clonality:
    Monoclonal
  • Clone:
    PCRP-KLF12-1E3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9Y4X4
  • Antigen synonyms:
    AP-2rep transcription factor|AP-2rep|AP2REP|AI225908|B130052C06Rik|D530033K05Rik|AP-2 repressor|AI315654|HSPC122
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    44,24 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human KLF12 protein.
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [PCRP-KLF12-1E3] antibody to KLF12 for IP, Flow Cytometry and IF with samples derived from human.

  • Validated Applications: IP, Flow Cytometry, IF
  • Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: KLF12
Clonality: Monoclonal
Clone: PCRP-KLF12-1E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human