Order Entry
Sweden
ContactUsLinkComponent
Anti-CYP24A1 Rabbit Polyclonal Antibody
Anti-CYP24A1 Rabbit Polyclonal Antibody
  ANTIA96276-100
New Product
undefined
Anti-CYP24A1 Rabbit Polyclonal Antibody
  ANTIA96276-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    1, 1 25 @dihydroxyvitamin D3 24 hydroxylase
  • Antigen symbol:
    CYP24A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q07973
  • Antigen synonyms:
    24-OHase|1 25 dihydroxyvitamin D(3) 24 hydroxylase|CP24|25-dihydroxyvitamin D(3) 24-hydroxylase|24 OHase|CP24A_HUMAN|CP 24|1,25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    58 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human Cytochrome P450 24A1 (amino acids 411-460)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to Cytochrome P450 24A1 for WB and ELISA with samples derived from Human.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: CYP24A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human