Order Entry
Sweden
ContactUsLinkComponent
 
Anti-TCEAL3 + TCEAL5 + TCEAL6 Rabbit Polyclonal Antibody
  ANTIA95272-100
New Product
undefined
Anti-TCEAL3 + TCEAL5 + TCEAL6 Rabbit Polyclonal Antibody
  ANTIA95272-100
New Product

 

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q969E4, Q5H9L2, Q6IPX3
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    22 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human TCEAL3 (amino acids 141-190)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to TCEAL3 + TCEAL5 + TCEAL6 for IHC and ELISA with samples derived from Human.

  • Validated applications: IHC, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human