Anti-NG2 Rabbit Polyclonal Antibody
Catalog # ANTIA309698-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:NG2
- Antigen symbol:NG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q8VHY0
- Antigen synonyms:Chondroitin sulfate proteoglycan NG2|Chondroitin sulfate proteoglycan 4 (melanoma-associated)|Cspg4 chondroitin sulfate proteoglycan 4|AN2 proteoglycan|Chondroitin sulfate proteoglycan 4|Cspg4|CSPG4_HUMAN|AN2|HMW-MAA|4732461B14Rik
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
- Molecular weight:270 - 300 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of human NG2/CSPG4.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to NG2 for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: NG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat