Anti-SLC12A1/NKCC2 Rabbit Monoclonal Antibody [clone: ARC2915]
Catalog # ANTIA309209-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:SLC12A1 / NKCC2
- Antigen symbol:SLC12A1 / NKCC2
- Clonality:Monoclonal
- Clone:ARC2915
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q13621
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:100 - 290 kDa
- Sequence:YGIYNMWVSLFGAVLCCAVMFVINWWAAVITYVIEFFLYVYVTCKKPDVNWGSSTQALSYVSALDNALELTTVEDHVKNFRPQCIVLTGGPMTRPALLDITHAFTKNSGLCICCEVFVGPRKLCVKEMNSGMAKKQAWLIKNKIKAFYAAVAADCFRDGVRSLLQASGLGRMKPNTLVIGYKKNWRKAPLTEIENYVGII
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 601-800 of human SLC12A1 / NKCC2 (Q13621).
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC2915] antibody to SLC12A1/NKCC2 for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: SLC12A1 / NKCC2
Clonality: Monoclonal
Clone: ARC2915
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat