Anti-CD63 Rabbit Recombinant Antibody [clone: LAMP3/8705R]
Catalog # ANTIA314729-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 63
- Antigen symbol:CD63
- Clonality:Recombinant
- Clone:LAMP3/8705R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08962
- Antigen synonyms:CD63_HUMAN|gp55|CD63|CD63 molecule|Granulophysin|CD63 antigen (melanoma 1 antigen)|CD63 antigen|LAMP 3|CD63 antigen melanoma 1 antigen
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:26 kDa (core protein)/30-60 kDa (glycosylated)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CD63 protein (recognizes extracellular epitope)
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [LAMP3/8705R] antibody to CD63 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: CD63
Clonality: Recombinant
Clone: LAMP3/8705R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human