Order Entry
Sweden
ContactUsLinkComponent
Anti-SynGAP Rabbit Monoclonal Antibody [clone: ARC2183]
Anti-SynGAP Rabbit Monoclonal Antibody [clone: ARC2183]
Katalog # ANTIA308630-100
Leverantör:  ANTIBODIES.COM
New Product
undefined
Anti-SynGAP Rabbit Monoclonal Antibody [clone: ARC2183]
Katalog # ANTIA308630-100
Leverantör:  ANTIBODIES.COM
New Product

Specifikationer

  • Antibody type:
    Primary
  • Antigen name:
    DKFZp761G1421
  • Antigen symbol:
    SynGAP
  • Clonality:
    Monoclonal
  • Clone:
    ARC2183
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q96PV0
  • Antigen synonyms:
    Ras GTPase-activating protein SynGAP|MRD5|Neuronal RasGAP|OTTHUMP00000064825|p135 SynGAP|Ras GTPase activating protein SynGAP|RASA 1|KIAA1938|RASA 5
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    140 kDa
  • Sequence:
    SEKRLRQQQAEKDSQIKSIIGRLMLVEEELRRDHPAMAEPLPEPKKRLLDAQERQLPPLGPTNPRVTLAPPWNGLAPPAPPPPPRLQITENGEFRNTADH
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 1244-1343 of human SynGAP (Q96PV0)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifikationer

Om denna produkt

Rabbit monoclonal [ARC2183] antibody to SynGAP for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2,000

Type: Primary
Antigen: SynGAP
Clonality: Monoclonal
Clone: ARC2183
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat