Anti-COASY Rabbit Monoclonal Antibody [clone: ARC2552]
Catalog # ANTIA308578-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Bifunctional coenzyme A synthase
- Antigen symbol:COASY
- Clonality:Monoclonal
- Clone:ARC2552
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q13057
- Antigen synonyms:Bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase|COASY|DPCOAK|Dephospho-CoA pyrophosphorylase|CoA synthase|COASY_HUMAN|DPCK|Dephospho-CoA kinase|Dephosphocoenzyme A kinase
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:62 kDa
- Sequence:RVCVIDAAVLLEAGWQNLVHEVWTAVIPETEAVRRIVERDGLSEAAAQSRLQSQMSGQQLVEQSHVVLSTLWEPHITQRQVEKAWALLQKRIPKTHQALD
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 465-564 of human CoA synthase (COASY) (Q13057)
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC2552] antibody to COASY for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: COASY
Clonality: Monoclonal
Clone: ARC2552
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human