Anti-Beclin 1 (phospho Ser15) Rabbit Polyclonal Antibody
Catalog # ANTIA308398-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:APG6
- Antigen symbol:Beclin 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q14457
- Antigen synonyms:Beclin 1 (coiled coil moesin like BCL2 interacting protein)|ATG6|ATG6 autophagy related 6 homolog|Beclin1|Beclin 1|Beclin-1|ATG 6|Beclin 1 autophagy related|Bcl-2-interacting protein beclin
- Modification:phospho Ser15
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
- Molecular weight:60 kDa
- Sequence:QVSFV
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic phosphorylated peptide around S15 of human Beclin 1 (NP_003757.1)
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to Beclin 1 (phospho Ser15) for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:100 to 1:500
Type: Primary
Antigen: Beclin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat