Anti-Histone H3 (di methyl Lys79) Rabbit Polyclonal Antibody
Catalog # ANTIA308295-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cit Histone H3
- Antigen symbol:Histone H3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- ChIP:Yes
- Form:Liquid
- Gene ID:UniprotID# Q16695
- Antigen synonyms:H3 3 like sequence MH921|FLJ92264|H3 a|citrullin|H3 3A|citrulline|Citrullated|H 3|Citrullinated Histone H3
- Modification:di methyl Lys79
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
- Molecular weight:17 kDa
- Sequence:QDFKT
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic dimethylated peptide around K79 of human histone H3 (NP_003520.1)
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to Histone H3 (di methyl Lys79) for WB, IP and ChIP with samples derived from human, mouse and rat.
- Validated applications: WB, IP, ChIP
- Recommended dilutions: WB: 1:100 to 1:500, ChIP: 1:50 to 1:200
Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat