Anti-CD48 Rabbit Recombinant Antibody [clone: DM44]
New Product
Recombinant rabbit monoclonal [DM44] antibody to CD48 for ELISA and flow cytometry with samples derived from human.
- Validated applications: ELISA, flow cytometry
- Recommended dilutions: ELISA: 1:5000 to 10000, flow cytometry: 1:100
Type: Primary
Antigen: CD48
Clonality: Recombinant
Clone: DM44
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ANTIA318651-500
- ANTIA318651-100
- Antigen symbol:
- CD48
- CD48
- Antigen name:
- Antigen CD48
- Antigen CD48
- Antigen synonyms:
- BCM1 surface antigen|B cell membrane protein|BLAST|BLAST1|BCM 1 surface antigen|BCM1|B lymphocyte activation marker BLAST 1|B-lymphocyte activation marker BLAST-1|B-cell activation marker
- BCM1 surface antigen|B cell membrane protein|BLAST|BLAST1|BCM 1 surface antigen|BCM1|B lymphocyte activation marker BLAST 1|B-lymphocyte activation marker BLAST-1|B-cell activation marker
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DM44
- DM44
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# P09326
- UniprotID# P09326
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilised
- Lyophilised
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Storage temperature:
- Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C