Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-CK14]
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Mouse monoclonal (ABT-CK14) antibody to Cytokeratin 14 for IHC and WB with samples derived from Human.
- Validated Applications: IHC, WB
Type: Primary
Antigen: p63 Protein + HMW Cytokeratin
Clonality: Monoclonal
Clone: ABT-CK14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ANTIA95696-50
- ANTIA95696-100
- Antigensymbol:
- p63 Protein + HMW Cytokeratin
- p63 Protein + HMW Cytokeratin
- Antikroppsnamn:
- CK13
- CK13
- Antigen synonymer:
- Cytokeratin 13|KRT13|Cytokeratin 15|KRT14|Cytokeratin 14|Cytokeratin 16|CK16|CK14|CK15
- Cytokeratin 13|KRT13|Cytokeratin 15|KRT14|Cytokeratin 14|Cytokeratin 16|CK16|CK14|CK15
- Antikroppstyp:
- Primär
- Primär
- Klonalitet:
- Monoclonal
- Monoclonal
- Konjugation:
- Unconjugated
- Unconjugated
- Klon:
- ABT-CK14
- ABT-CK14
- Reaktivitet:
- Human
- Human
- Värd:
- Mouse
- Mouse
- Gen-ID:
- UniprotID# P02533
- UniprotID# P02533
- Isotyp:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunKemi:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human Cytokeratin 14
- Synthetic peptide derived from human Cytokeratin 14
- Form:
- Liquid
- Liquid
- Rening:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage buffer:
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- IHC
- IHC