Anti-Androgen Receptor Mouse Monoclonal Antibody [clone: ABT-AR]
New Product
Mouse monoclonal (ABT-AR) antibody to Androgen Receptor for IHC and WB with samples derived from Human.
- Validated Applications: IHC, WB
Type: Primary
Antigen: Androgen Receptor
Clonality: Monoclonal
Clone: ABT-AR
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
- Catalog No:
- ANTIA95527-100
- ANTIA95527-50
- Antigen symbol:
- Androgen Receptor
- Androgen Receptor
- Antigen name:
- AIS
- AIS
- Antigen synonyms:
- Androgen Receptor| Kennedy disease)|Androgen receptor (dihydrotestosterone receptor| testicular feminization|AR|ANDR_HUMAN|androgen receptor splice variant 4b|Androgen nuclear receptor variant 2| spinal and bulbar muscular atrophy
- Androgen Receptor| Kennedy disease)|Androgen receptor (dihydrotestosterone receptor| testicular feminization|AR|ANDR_HUMAN|androgen receptor splice variant 4b|Androgen nuclear receptor variant 2| spinal and bulbar muscular atrophy
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-AR
- ABT-AR
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P10275
- UniprotID# P10275
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human Androgen Receptor
- Synthetic peptide derived from human Androgen Receptor
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage buffer:
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- IHC
- IHC