Order Entry
Sweden
ContactUsLinkComponent
 
Anti-PRR7 Mouse Monoclonal Antibody [Clone: TRAP3/10]
  ANTIA85830-100
New Product
undefined
Anti-PRR7 Mouse Monoclonal Antibody [Clone: TRAP3/10]
  ANTIA85830-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Proline rich protein 7
  • Antigen symbol:
    PRR7
  • Clonality:
    Monoclonal
  • Clone:
    TRAP3/10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8TB68
  • Antigen synonyms:
    PRR7|Synaptic proline rich membrane protein|TRAP3|Proline-rich protein 7|transmembrane adaptor protein 3|Synaptic proline-rich membrane protein|PRR7_HUMAN
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253)
  • Tested applications:
    ICC
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [TRAP3/10] antibody to PRR7 for WB and ICC with samples derived from human, mouse and rat.

  • Validated applications: WB, ICC
  • Recommended dilutions: ICC: 10 µg/ml, WB: 1 µg/ml

Type: Primary
Antigen: PRR7
Clonality: Monoclonal
Clone: TRAP3/10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat