Anti-Npc2 Goat Polyclonal Antibody
Catalog # ANTIA84948-100
Leverantör: ANTIBODIES.COM
New Product
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:EDDM1
- Antigensymbol:Npc2
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Goat
- Isotyp:IgG
- Reaktivitet:Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonymer:Human epididymis-specific protein 1|Epididymal secretory protein|Epididymal secretory protein E1|Niemann-Pick disease type C2 protein|Niemann-Pick disease type C2|Epididymal protein 1|Niemann Pick C2|hE1|NPC intracellular cholesterol transporter 2
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:QAEPLHFKDCGSK
- Förvaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Koncentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Mouse Npc2 (internal region)
- Rening:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Förpackningstyp:Plastic vial
- Förp.:100 µG
Specifikationer
Om denna produkt
Goat polyclonal antibody to NPC2 for ELISA and WB with samples derived from mouse.
- Validated applications: ELISA, W
Type: Primary
Antigen: Npc2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse