Anti-Ku70 Goat Polyclonal Antibody
Catalog # ANTIA84357-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:5''-deoxyribose-5-phosphate lyase Ku70
- Antigen symbol:Ku70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CTC box binding factor 75 kDa subunit|ATP dependent DNA helicase II 70 kDa subunit|70 kDa subunit of Ku antigen|CTC75|ATP dependent DNA helicase 2 subunit 1|CTC box-binding factor 75 kDa subunit|ATP-dependent DNA helicase 2 subunit 1|5''-dRP lyase Ku70|ATP-dependent DNA helicase II 70 kDa subunit
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:YKTEGDEEAEEEQ-C
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human XRCC6 (N terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to KU70 for ELISA and WB with samples derived from human.
- Validated applications: ELISA, W
Type: Primary
Antigen: Ku70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human