Anti-TBP like protein TLP Goat Polyclonal Antibody
ANTIA83875-100
New Product
- Antibody type:Primary
- Antigen name:21 kDa TBP-like protein
- Antigen symbol:TBP like protein TLP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TATA box binding protein-related factor 2|STUD|TATA box-binding protein-like protein 1|TATA box-binding protein-related factor 2|TATA box binding protein like protein 1|Second TBP of unique DNA|Second TBP of unique DNA protein|21kDa TBP like protein|TBP like 1
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-QIYPFVFESRKEIL
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human TBPL1 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to TBP like protein TLP for ELISA and WB with samples derived from human.
- Validated applications: ELISA, W
Type: Primary
Antigen: TBP like protein TLP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human