Anti-AKAP3 Rabbit Polyclonal Antibody
Catalog # ANTIA99180-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:A kinase (PRKA) anchor protein 3
- Antigensymbol:AKAP3
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Human,Mouse
- Form:Liquid
- Gen-ID:UniprotID# O75969
- Antigen synonymer:AKAP-3|AKAP 110|A-kinase anchor protein 110 kDa|AKAP3|A-kinase anchor protein 3|Cancer/testis antigen 82|AKAP110|AKAP3_HUMAN|CT82
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molekylvikt:94 kDa
- Förvaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human AKAP3 (amino acids 191-240)
- Rening:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Förpackningstyp:Plastic vial
- Förp.:100 µG
Specifications
About this item
Rabbit polyclonal antibody to AKAP3 for IF and ELISA with samples derived from human and mouse.
- Validated applications: IF, ELISA
- Recommended dilutions: IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: AKAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse