Order Entry
Sweden
ContactUsLinkComponent
Anti-CD11a Rabbit Monoclonal Antibody [clone: ARC0344]
Anti-CD11a Rabbit Monoclonal Antibody [clone: ARC0344]
Catalog # ANTIA308023-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CD11a Rabbit Monoclonal Antibody [clone: ARC0344]
Catalog # ANTIA308023-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen CD11A
  • Antigen symbol:
    CD11a
  • Clonality:
    Monoclonal
  • Clone:
    ARC0344
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P20701
  • Antigen synonyms:
    CD 11a|CD11a antigen|alpha polypeptide)|Integrin, alpha L (antigen CD11A (p180), lymphocyte function associated antigen 1|Integrin gene promoter|Antigen CD11A (p180), lymphocyte function associated antigen 1, alpha polypeptide|Integrin alpha-L|CD11 antigen-like family member A|Integrin Alpha L
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    180 kDa
  • Sequence:
    NASLAQVVMKVDVVYEKQMLYLYVLSGIGGLLLLLLIFIVLYKVGFFKRNLKEKMEAGRGVPNGIPAEDSEQLASGQEAGDPGCLKPLHEKDSESGGGKD
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 1071-1170 of human CD11a/LFA-1A/ITGAL (P20701)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC0344] antibody to CD11a for WB and IHC with samples derived from Human.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone: ARC0344
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human