Order Entry
Sweden
ContactUsLinkComponent
 
Anti-FOXO3A Goat Polyclonal Antibody
  ANTIA83729-100
New Product
undefined
Anti-FOXO3A Goat Polyclonal Antibody
  ANTIA83729-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    AF6q21
  • Antigen symbol:
    FOXO3A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    Forkhead box O3|FKHRL1P2|DKFZp781A0677|Forkhead box O3A|FKHRL 1|FKHRL1|Forkhead (Drosophila) homolog (rhabdomyosarcoma) like 1|AF6q21 protein|FKHR2
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
  • Sequence:
    C-GAKQASSQSWVPG
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human FOXO3 (C terminal)
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Goat polyclonal antibody to FOXO3A for ELISA and WB with samples derived from human.

  • Validated applications: ELISA, W

Type: Primary
Antigen: FOXO3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human