Anti-WTAP Mouse Monoclonal Antibody [clone: PCRP-WTAP-1A4]
Catalog # ANTIA317179-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:DKFZp686F20131
- Antigen symbol:WTAP
- Clonality:Monoclonal
- Clone:PCRP-WTAP-1A4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q15007
- Antigen synonyms:OTTHUMP00000017523|FL2D_HUMAN|hFL(2)D|Female-lethal(2)D homolog|PNAS 132|OTTHUMP00000017522|MGC3925|KIAA0105|Mum2
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without Sodium azide and carrier free
- Molecular weight:44,2 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human WTAP protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PCRP-WTAP-1A4] antibody to WTAP for flow cytometry and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: WTAP
Clonality: Monoclonal
Clone: PCRP-WTAP-1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human