Order Entry
Sweden
ContactUsLinkComponent
Anti-TL1A Rabbit Recombinant Antibody [clone: VEGI/7799R]
Anti-TL1A Rabbit Recombinant Antibody [clone: VEGI/7799R]
Catalog # ANTIA317118-100
Supplier:  ANTIBODIES.COM
New Product
Anti-TL1A Rabbit Recombinant Antibody [clone: VEGI/7799R]
Catalog # ANTIA317118-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    MGC129934
  • Antigensymbol:
    TL1A
  • Klonalitet:
    Recombinant
  • Klon:
    VEGI/7799R
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG kappa
  • Reaktivitet:
    Human
  • Form:
    Liquid
  • Gen-ID:
    UniprotID# O95150
  • Antigen synonymer:
    TNF ligand-related molecule 1|TL1A|tl1|tnf superfamily ligand tl1a|TNFSF15|TL1 A|tnf ligand related molecule 1|TNF15_HUMAN|MGC129935
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without Sodium azide and carrier free
  • Molekylvikt:
    22 kDa
  • Förvaringstemperatur:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Koncentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human VEGI protein.
  • Tested applications:
    IHC-P
  • Rening:
    Protein A chromatography
  • Förpackningstyp:
    Plastic vial
  • Förp.:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [VEGI/7799R] antibody to TL1A for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: TL1A
Clonality: Recombinant
Clone: VEGI/7799R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human