Anti-BCMA Recombinant Recombinant Antibody
Catalog # ANTIA318928-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:B cell maturation antigen
- Antigen symbol:BCMA
- Clonality:Recombinant
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Recombinant
- Isotype:IgG1
- Reactivity:Human
- Form:Lyophilized
- Gene ID:UniprotID# Q02223
- Antigen synonyms:B cell maturation factor|CD269|B cell maturation protein|TNFRSF17|TNR17_HUMAN|B-cell maturation protein|BCMA|BCM|CD269 antigen
- Storage buffer:Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Storage temperature:Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Concentration:Reconstitution dependent
- Shipping temperature:Shipped on blue ice at 4 °C
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant humanized monoclonal [Belantamab Biosimilar] antibody to BCMA for ELISA with samples derived from human.
- Validated applications: ELISA
- Recommended dilutions: ELISA: 1:5000 to 10000
Type: Primary
Antigen: BCMA
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human